RIE156328079 Rieke Metals, Inc.

3-(2,3-dimethylphenyl)-4'-fluoropropiophenone

  • MDL number MFCD03843627
  • Cas Number : 898769-50-1
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2,3-dimethylphenyl)-1-(4-fluorophenyl)-1-propanone
Inchi Code 1S/C17H17FO/c1-12-4-3-5-14(13(12)2)8-11-17(19)15-6-9-16(18)10-7-15/h3-7,9-10H,8,11H2,1-2H3
Inchi Key GHXKFZGXTNYLFL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2600-23

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-23
Availability details for
Quantity
  • Availability
  • Add to Cart