RIE156328086 Rieke Metals, Inc.

4'-bromo-3-(2,3-dimethylphenyl)-3'-fluoropropiophenone

  • MDL number MFCD03843634
  • Cas Number : 898792-89-7
  • Molecular Weight : 335.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromo-3-fluorophenyl)-3-(2,3-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16BrFO/c1-11-4-3-5-13(12(11)2)7-9-17(20)14-6-8-15(18)16(19)10-14/h3-6,8,10H,7,9H2,1-2H3
Inchi Key KSWLLTHFDMKKGP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2600-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-32
Availability details for
Quantity
  • Availability
  • Add to Cart