RIE156328087 Rieke Metals, Inc.

4'-chloro-3-(2,3-dimethylphenyl)-3'-fluoropropiophenone

  • MDL number MFCD03843635
  • Cas Number : 898792-92-2
  • Molecular Weight : 290.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2600-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-33
Availability details for
Quantity
  • Availability
  • Add to Cart