RIE156328088 Rieke Metals, Inc.

3'-chloro-3-(2,3-dimethylphenyl)-4'-fluoropropiophenone

  • MDL number MFCD03843636
  • Cas Number : 898792-95-5
  • Molecular Weight : 290.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-chloro-4-fluorophenyl)-3-(2,3-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16ClFO/c1-11-4-3-5-13(12(11)2)7-9-17(20)14-6-8-16(19)15(18)10-14/h3-6,8,10H,7,9H2,1-2H3
Inchi Key VHRHVEAYSSLUIW-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2600-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-34
Availability details for
Quantity
  • Availability
  • Add to Cart