RIE156328089 Rieke Metals, Inc.

2'-chloro-3-(2,3-dimethylphenyl)propiophenone

  • MDL number MFCD03843637
  • Cas Number : 898792-98-8
  • Molecular Weight : 272.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2600-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-36
Availability details for
Quantity
  • Availability
  • Add to Cart