RIE156328091 Rieke Metals, Inc.

3-(2,3-dimethylphenyl)-2'-trifluoromethylpropiophenone

  • MDL number MFCD03843639
  • Cas Number : 898793-04-9
  • Molecular Weight : 306.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2600-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-43
Availability details for
Quantity
  • Availability
  • Add to Cart