RIE156328095 Rieke Metals, Inc.

2'-chloro-3-(2,3-dimethylphenyl)-4'-fluoropropiophenone

  • MDL number MFCD03843643
  • Cas Number : 898793-16-3
  • Molecular Weight : 290.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2600-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-50
Availability details for
Quantity
  • Availability
  • Add to Cart