RIE156328098 Rieke Metals, Inc.

2',3'-dichloro-3-(2,3-dimethylphenyl)propiophenone

  • MDL number MFCD03843646
  • Cas Number : 898793-23-2
  • Molecular Weight : 307.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2600-53

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-53
Availability details for
Quantity
  • Availability
  • Add to Cart