RIE156328102 Rieke Metals, Inc.

3',5'-dichloro-3-(2,3-dimethylphenyl)propiophenone

  • MDL number MFCD03843650
  • Cas Number : 898793-31-2
  • Molecular Weight : 307.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3,5-dichlorophenyl)-3-(2,3-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16Cl2O/c1-11-4-3-5-13(12(11)2)6-7-17(20)14-8-15(18)10-16(19)9-14/h3-5,8-10H,6-7H2,1-2H3
Inchi Key SPOGHIYVBIEART-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2600-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-57
Availability details for
Quantity
  • Availability
  • Add to Cart