RIE156328103 Rieke Metals, Inc.

2',4'-difluoro-3-(2,3-dimethylphenyl)propiophenone

  • MDL number MFCD03843651
  • Cas Number : 898793-33-4
  • Molecular Weight : 274.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,4-difluorophenyl)-3-(2,3-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16F2O/c1-11-4-3-5-13(12(11)2)6-9-17(20)15-8-7-14(18)10-16(15)19/h3-5,7-8,10H,6,9H2,1-2H3
Inchi Key NUFVDJCNAWVJRS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2600-58

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-58
Availability details for
Quantity
  • Availability
  • Add to Cart