RIE156328106 Rieke Metals, Inc.

3-(2,3-dimethylphenyl)-3',4',5'-trifluoropropiophenone

  • MDL number MFCD03843654
  • Cas Number : 898793-39-0
  • Molecular Weight : 292.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2600-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2600-61
Availability details for
Quantity
  • Availability
  • Add to Cart