RIE156328108 Rieke Metals, Inc.

cyclopropyl 2-(2,3-dimethylphenyl)ethyl ketone

  • MDL number MFCD03843655
  • Cas Number : 898793-43-6
  • Molecular Weight : 202.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2601-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2601-00
Availability details for
Quantity
  • Availability
  • Add to Cart