RIE156328109 Rieke Metals, Inc.

cyclobutyl 2-(2,3-dimethylphenyl)ethyl ketone

  • MDL number MFCD03843656
  • Cas Number : 898793-45-8
  • Molecular Weight : 216.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2602-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2602-00
Availability details for
Quantity
  • Availability
  • Add to Cart