RIE156328110 Rieke Metals, Inc.

cyclopentyl 2-(2,3-dimethylphenyl)ethyl ketone

  • MDL number MFCD03843657
  • Cas Number : 898793-47-0
  • Molecular Weight : 230.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclopentyl-3-(2,3-dimethylphenyl)-1-propanone
Inchi Code 1S/C16H22O/c1-12-6-5-9-14(13(12)2)10-11-16(17)15-7-3-4-8-15/h5-6,9,15H,3-4,7-8,10-11H2,1-2H3
Inchi Key DWXSFFHIIVRILK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2603-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2603-00
Availability details for
Quantity
  • Availability
  • Add to Cart