RIE156328113 Rieke Metals, Inc.

3-(2,4-dimethylphenyl)-2'-methylpropiophenone

  • MDL number MFCD03843660
  • Cas Number : 898793-53-8
  • Molecular Weight : 252.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2700-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2700-02
Availability details for
Quantity
  • Availability
  • Add to Cart