RIE156328116 Rieke Metals, Inc.

3-(2,4-dimethylphenyl)-2'-methoxypropiophenone

  • MDL number MFCD03843663
  • Cas Number : 898793-59-4
  • Molecular Weight : 268.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2700-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2700-05
Availability details for
Quantity
  • Availability
  • Add to Cart