RIE156328122 Rieke Metals, Inc.

2'-carboethoxy-3-(2,4-dimethylphenyl)propiophenone

  • MDL number MFCD03843669
  • Cas Number : 898793-71-0
  • Molecular Weight : 310.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-[3-(2,4-dimethylphenyl)propanoyl]benzoate
Inchi Code 1S/C20H22O3/c1-4-23-20(22)18-8-6-5-7-17(18)19(21)12-11-16-10-9-14(2)13-15(16)3/h5-10,13H,4,11-12H2,1-3H3
Inchi Key ODFYFIDCAPZHDR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2700-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2700-11
Availability details for
Quantity
  • Availability
  • Add to Cart