RIE156328131 Rieke Metals, Inc.

3-(2,4-dimethylphenyl)-3'-fluoropropiophenone

  • MDL number MFCD03843678
  • Cas Number : 898793-96-9
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2700-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2700-22
Availability details for
Quantity
  • Availability
  • Add to Cart