RIE156328140 Rieke Metals, Inc.

4'-chloro-3-(2,4-dimethylphenyl)-3'-fluoropropiophenone

  • MDL number MFCD03843687
  • Cas Number : 898794-22-4
  • Molecular Weight : 290.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-chloro-3-fluorophenyl)-3-(2,4-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16ClFO/c1-11-3-4-13(12(2)9-11)6-8-17(20)14-5-7-15(18)16(19)10-14/h3-5,7,9-10H,6,8H2,1-2H3
Inchi Key KHFQEPSTYJDQBA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2700-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2700-33
Availability details for
Quantity
  • Availability
  • Add to Cart