RIE156328141 Rieke Metals, Inc.

3'-chloro-3-(2,4-dimethylphenyl)-4'-fluoropropiophenone

  • MDL number MFCD03843688
  • Cas Number : 898794-24-6
  • Molecular Weight : 290.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-chloro-4-fluorophenyl)-3-(2,4-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16ClFO/c1-11-3-4-13(12(2)9-11)6-8-17(20)14-5-7-16(19)15(18)10-14/h3-5,7,9-10H,6,8H2,1-2H3
Inchi Key OHRGMRRGLPTTAB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2700-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2700-34
Availability details for
Quantity
  • Availability
  • Add to Cart