RIE156328142 Rieke Metals, Inc.

2'-chloro-3-(2,4-dimethylphenyl)propiophenone

  • MDL number MFCD03843689
  • Cas Number : 898794-26-8
  • Molecular Weight : 272.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-chlorophenyl)-3-(2,4-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H17ClO/c1-12-7-8-14(13(2)11-12)9-10-17(19)15-5-3-4-6-16(15)18/h3-8,11H,9-10H2,1-2H3
Inchi Key MXAOZSUIJOFPEG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2700-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2700-36
Availability details for
Quantity
  • Availability
  • Add to Cart