RIE156328146 Rieke Metals, Inc.

3-(2,4-dimethylphenyl)-4'-trifluoromethylpropiophenone

  • MDL number MFCD03843693
  • Cas Number : 898794-34-8
  • Molecular Weight : 306.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2,4-dimethylphenyl)-1-[4-(trifluoromethyl)phenyl]-1-propanone
Inchi Code 1S/C18H17F3O/c1-12-3-4-14(13(2)11-12)7-10-17(22)15-5-8-16(9-6-15)18(19,20)21/h3-6,8-9,11H,7,10H2,1-2H3
Inchi Key XJKXTXRTNUAYPZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2700-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2700-45
Availability details for
Quantity
  • Availability
  • Add to Cart