RIE156328149 Rieke Metals, Inc.

3'-chloro-3-(2,4-dimethylphenyl)-5'-fluoropropiophenone

  • MDL number MFCD03843696
  • Cas Number : 898794-40-6
  • Molecular Weight : 290.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2700-51

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2700-51
Availability details for
Quantity
  • Availability
  • Add to Cart