RIE156328155 Rieke Metals, Inc.

3',5'-dichloro-3-(2,4-dimethylphenyl)propiophenone

  • MDL number MFCD03843702
  • Cas Number : 898794-52-0
  • Molecular Weight : 307.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3,5-dichlorophenyl)-3-(2,4-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16Cl2O/c1-11-3-4-13(12(2)7-11)5-6-17(20)14-8-15(18)10-16(19)9-14/h3-4,7-10H,5-6H2,1-2H3
Inchi Key IERWXHGOCKIXFL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2700-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2700-57
Availability details for
Quantity
  • Availability
  • Add to Cart