RIE156328164 Rieke Metals, Inc.

cyclohexyl 2-(2,4-dimethylphenyl)ethyl ketone

  • MDL number MFCD03843710
  • Cas Number : 898794-70-2
  • Molecular Weight : 244.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2704-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2704-00
Availability details for
Quantity
  • Availability
  • Add to Cart