RIE156328168 Rieke Metals, Inc.

3-(2,5-dimethylphenyl)-4'-methylpropiophenone

  • MDL number MFCD03843714
  • Cas Number : 898794-78-0
  • Molecular Weight : 252.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2800-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2800-04
Availability details for
Quantity
  • Availability
  • Add to Cart