RIE156328171 Rieke Metals, Inc.

3-(2,5-dimethylphenyl)-4'-methoxypropiophenone

  • MDL number MFCD03843717
  • Cas Number : 898794-84-8
  • Molecular Weight : 268.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2800-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2800-07
Availability details for
Quantity
  • Availability
  • Add to Cart