RIE156328173 Rieke Metals, Inc.

3'-cyano-3-(2,5-dimethylphenyl)propiophenone

  • MDL number MFCD03843719
  • Cas Number : 898794-88-2
  • Molecular Weight : 263.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2800-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2800-09
Availability details for
Quantity
  • Availability
  • Add to Cart