RIE156328176 Rieke Metals, Inc.

3'-carboethoxy-3-(2,5-dimethylphenyl)propiophenone

  • MDL number MFCD03843722
  • Cas Number : 898794-94-0
  • Molecular Weight : 310.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2800-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2800-12
Availability details for
Quantity
  • Availability
  • Add to Cart