RIE156328177 Rieke Metals, Inc.

4'-carboethoxy-3-(2,5-dimethylphenyl)propiophenone

  • MDL number MFCD03843723
  • Cas Number : 898794-96-2
  • Molecular Weight : 310.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-[3-(2,5-dimethylphenyl)propanoyl]benzoate
Inchi Code 1S/C20H22O3/c1-4-23-20(22)17-9-7-16(8-10-17)19(21)12-11-18-13-14(2)5-6-15(18)3/h5-10,13H,4,11-12H2,1-3H3
Inchi Key JAYAIZJCFGYHOU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2800-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2800-13
Availability details for
Quantity
  • Availability
  • Add to Cart