RIE156328183 Rieke Metals, Inc.

4'-chloro-3-(2,5-dimethylphenyl)propiophenone

  • MDL number MFCD03843729
  • Cas Number : 898753-39-4
  • Molecular Weight : 272.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-chlorophenyl)-3-(2,5-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H17ClO/c1-12-3-4-13(2)15(11-12)7-10-17(19)14-5-8-16(18)9-6-14/h3-6,8-9,11H,7,10H2,1-2H3
Inchi Key GIEZBHZHQHFROA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2800-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2800-21
Availability details for
Quantity
  • Availability
  • Add to Cart