RIE156328187 Rieke Metals, Inc.

2',4'-dimethyl-3-(2,5-dimethylphenyl)propiophenone

  • MDL number MFCD03843733
  • Cas Number : 898753-51-0
  • Molecular Weight : 266.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2800-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2800-27
Availability details for
Quantity
  • Availability
  • Add to Cart