RIE156328189 Rieke Metals, Inc.

2',6'-dimethyl-3-(2,5-dimethylphenyl)propiophenone

  • MDL number MFCD03843735
  • Cas Number : 898753-57-6
  • Molecular Weight : 266.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2800-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2800-29
Availability details for
Quantity
  • Availability
  • Add to Cart