RIE156328191 Rieke Metals, Inc.

3',5'-dimethyl-3-(2,5-dimethylphenyl)propiophenone

  • MDL number MFCD03843737
  • Cas Number : 898753-63-4
  • Molecular Weight : 266.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2,5-dimethylphenyl)-1-(3,5-dimethylphenyl)-1-propanone
Inchi Code 1S/C19H22O/c1-13-5-6-16(4)17(10-13)7-8-19(20)18-11-14(2)9-15(3)12-18/h5-6,9-12H,7-8H2,1-4H3
Inchi Key UCQGEVVOCQOXDK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2800-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2800-31
Availability details for
Quantity
  • Availability
  • Add to Cart