RIE156328197 Rieke Metals, Inc.

3-(2,5-dimethylphenyl)-2'-trifluoromethylpropiophenone

  • MDL number MFCD03843743
  • Cas Number : 898753-81-6
  • Molecular Weight : 306.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2800-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2800-43
Availability details for
Quantity
  • Availability
  • Add to Cart