RIE156328198 Rieke Metals, Inc.

3-(2,5-dimethylphenyl)-3'-trifluoromethylpropiophenone

  • MDL number MFCD03843744
  • Cas Number : 898753-84-9
  • Molecular Weight : 306.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2800-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2800-44
Availability details for
Quantity
  • Availability
  • Add to Cart