RIE156328203 Rieke Metals, Inc.

4'-chloro-3-(2,5-dimethylphenyl)-2'-fluoropropiophenone

  • MDL number MFCD03843749
  • Cas Number : 898753-94-1
  • Molecular Weight : 290.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-chloro-2-fluorophenyl)-3-(2,5-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16ClFO/c1-11-3-4-12(2)13(9-11)5-8-17(20)15-7-6-14(18)10-16(15)19/h3-4,6-7,9-10H,5,8H2,1-2H3
Inchi Key DLHYNKBWWSAVSP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2800-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2800-52
Availability details for
Quantity
  • Availability
  • Add to Cart