RIE156328207 Rieke Metals, Inc.

3',4'-dichloro-3-(2,5-dimethylphenyl)propiophenone

  • MDL number MFCD03843753
  • Cas Number : 898754-02-4
  • Molecular Weight : 307.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3,4-dichlorophenyl)-3-(2,5-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H16Cl2O/c1-11-3-4-12(2)13(9-11)6-8-17(20)14-5-7-15(18)16(19)10-14/h3-5,7,9-10H,6,8H2,1-2H3
Inchi Key KLRRVFKALVNBBS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2800-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2800-56
Availability details for
Quantity
  • Availability
  • Add to Cart