RIE156328217 Rieke Metals, Inc.

cyclohexyl 2-(2,5-dimethylphenyl)ethyl ketone

  • MDL number MFCD03843762
  • Cas Number : 898754-22-8
  • Molecular Weight : 244.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclohexyl-3-(2,5-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H24O/c1-13-8-9-14(2)16(12-13)10-11-17(18)15-6-4-3-5-7-15/h8-9,12,15H,3-7,10-11H2,1-2H3
Inchi Key MSMCRAWBJAIJCZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2804-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2804-00
Availability details for
Quantity
  • Availability
  • Add to Cart