RIE156328223 Rieke Metals, Inc.

3-(2,6-dimethylphenyl)-3'-methoxypropiophenone

  • MDL number MFCD03843768
  • Cas Number : 898754-34-2
  • Molecular Weight : 268.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2900-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-06
Availability details for
Quantity
  • Availability
  • Add to Cart