RIE156328226 Rieke Metals, Inc.

3'-cyano-3-(2,6-dimethylphenyl)propiophenone

  • MDL number MFCD03843771
  • Cas Number : 898754-43-3
  • Molecular Weight : 263.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2900-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-09
Availability details for
Quantity
  • Availability
  • Add to Cart