RIE156328227 Rieke Metals, Inc.

4'-cyano-3-(2,6-dimethylphenyl)propiophenone

  • MDL number MFCD03843772
  • Cas Number : 898754-46-6
  • Molecular Weight : 263.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[3-(2,6-dimethylphenyl)propanoyl]benzonitrile
Inchi Code 1S/C18H17NO/c1-13-4-3-5-14(2)17(13)10-11-18(20)16-8-6-15(12-19)7-9-16/h3-9H,10-11H2,1-2H3
Inchi Key QYAKRKHAEYTLKP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2900-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-10
Availability details for
Quantity
  • Availability
  • Add to Cart