RIE156328228 Rieke Metals, Inc.

2'-carboethoxy-3-(2,6-dimethylphenyl)propiophenone

  • MDL number MFCD03843773
  • Cas Number : 898754-49-9
  • Molecular Weight : 310.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2900-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-11
Availability details for
Quantity
  • Availability
  • Add to Cart