RIE156328229 Rieke Metals, Inc.

3'-carboethoxy-3-(2,6-dimethylphenyl)propiophenone

  • MDL number MFCD03843774
  • Cas Number : 898754-52-4
  • Molecular Weight : 310.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2900-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-12
Availability details for
Quantity
  • Availability
  • Add to Cart