RIE156328231 Rieke Metals, Inc.

3-(2,6-dimethylphenyl)-2'-thiomethylpropiophenone

  • MDL number MFCD03843776
  • Cas Number : 898754-58-0
  • Molecular Weight : 284.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2,6-dimethylphenyl)-1-[2-(methylsulfanyl)phenyl]-1-propanone
Inchi Code 1S/C18H20OS/c1-13-7-6-8-14(2)15(13)11-12-17(19)16-9-4-5-10-18(16)20-3/h4-10H,11-12H2,1-3H3
Inchi Key BEFDGZTYSSKCCG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2900-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-14
Availability details for
Quantity
  • Availability
  • Add to Cart