RIE156328234 Rieke Metals, Inc.

4'-bromo-3-(2,6-dimethylphenyl)propiophenone

  • MDL number MFCD03843779
  • Cas Number : 898754-67-1
  • Molecular Weight : 317.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromophenyl)-3-(2,6-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H17BrO/c1-12-4-3-5-13(2)16(12)10-11-17(19)14-6-8-15(18)9-7-14/h3-9H,10-11H2,1-2H3
Inchi Key ZUQKXOQYKGRVMF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2900-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-19
Availability details for
Quantity
  • Availability
  • Add to Cart