RIE156328235 Rieke Metals, Inc.

3'-chloro-3-(2,6-dimethylphenyl)propiophenone

  • MDL number MFCD03843780
  • Cas Number : 898754-70-6
  • Molecular Weight : 272.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-chlorophenyl)-3-(2,6-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H17ClO/c1-12-5-3-6-13(2)16(12)9-10-17(19)14-7-4-8-15(18)11-14/h3-8,11H,9-10H2,1-2H3
Inchi Key MVPFRZRQLSTZSL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2900-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-20
Availability details for
Quantity
  • Availability
  • Add to Cart