RIE156328237 Rieke Metals, Inc.

3-(2,6-dimethylphenyl)-3'-fluoropropiophenone

  • MDL number MFCD03843782
  • Cas Number : 898754-76-2
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2900-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-22
Availability details for
Quantity
  • Availability
  • Add to Cart