RIE156328238 Rieke Metals, Inc.

3-(2,6-dimethylphenyl)-4'-fluoropropiophenone

  • MDL number MFCD03843783
  • Cas Number : 898754-79-5
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2900-23

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-23
Availability details for
Quantity
  • Availability
  • Add to Cart