RIE156328240 Rieke Metals, Inc.

2',4'-dimethyl-3-(2,6-dimethylphenyl)propiophenone

  • MDL number MFCD03843785
  • Cas Number : 898754-84-2
  • Molecular Weight : 266.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,4-dimethylphenyl)-3-(2,6-dimethylphenyl)-1-propanone
Inchi Code 1S/C19H22O/c1-13-8-9-18(16(4)12-13)19(20)11-10-17-14(2)6-5-7-15(17)3/h5-9,12H,10-11H2,1-4H3
Inchi Key RKXNIDAZVHZGMK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2900-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2900-27
Availability details for
Quantity
  • Availability
  • Add to Cart